Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037523.1 | NP_001032612.1 | 1 - 2 | 111 | 77 | |
XM_006253375.2 | XP_006253437.1 | 2 - 3 | 235 | 77 | ||
GenBank mRNA | AY621362.1 | - | 1 - 2 | 111 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons |