Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037205.1 | NP_001032282.1 | 10 - 11 | 1299 | 129 | |
GenBank mRNA | BC100073.1 | - | 10 - 11 | 1299 | 7372627 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |