Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037205.1 | NP_001032282.1 | 4 - 5 | 455 | 139 | |
GenBank mRNA | BC100073.1 | - | 4 - 5 | 455 | 7372627 | 139 |
Gene Symbol: LOC103690063 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769965.1 | XP_008768187.1 | 4 - 5 | 1027 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |