Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35e4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153316.2 | NP_695228.2 | 2 | 1448 | 108 | |
GenBank mRNA | AF182714.1 | - | 3 | 1171 | 108 | |
BC085693.1 | - | 2 | 1448 | 7191218 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon |