Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145334.1 | NP_001138806.1 | 1 | 679 | 75 | |
GenBank mRNA | EU809461.1 | - | 1 | 679 | 75 | |
EU809462.1 | - | 1 | 655 | 75 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Both primers and probe map within a single exon |