Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008956.1 | NP_001008956.1 | 1 | 839 | 124 | |
GenBank mRNA | AY510314.1 | - | 1 | 839 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |