Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam26a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169119.1 | NP_001162590.1 | 83 | 82 | ||
GenBank mRNA | BC085958.1 | - | 83 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Protease 3' Most Mammalian Gene Collection Rattus norvegicus |