Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019171.1 | NP_062044.1 | 3 - 4 | 273 | 94 | |
GenBank mRNA | M33976.1 | - | 3 - 4 | 273 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons |