Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138890.2 | NP_620245.2 | 4 - 5 | 1312 | 102 | |
XM_006239774.2 | XP_006239836.1 | 5 - 6 | 1557 | 102 | ||
XM_008764503.1 | XP_008762725.1 | 4 - 5 | 1158 | 102 | ||
GenBank mRNA | AF494093.2 | - | 4 - 5 | 1312 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |