Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099461.1 | NP_001092931.1 | 2 - 3 | 490 | 100 | |
XM_008768512.1 | XP_008766734.1 | 2 - 3 | 1096 | 100 |
Gene Symbol: Vom2r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099460.1 | NP_001092930.1 | 484 | 100 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |