Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcgf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107245.2 | NP_001100715.2 | 4 - 5 | 429 | 71 | |
GenBank mRNA | BC167000.1 | - | 4 - 5 | 429 | 9031673 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |