Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcte3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191559.1 | NP_001178488.1 | 1 - 2 | 106 | 90 | |
XM_006227971.2 | XP_006228033.1 | 1 - 2 | 228 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Probe spans exons |