Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197217.1 | NP_001184146.1 | 1 - 2 | 277 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |