Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sohlh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191852.1 | NP_001178781.1 | 1 - 2 | 69 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |