Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Triml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191690.1 | NP_001178619.1 | 5 - 6 | 643 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |