Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013240.2 | NP_001013258.1 | 3 - 4 | 352 | 72 | |
NM_001267054.1 | NP_001253983.1 | 3 - 4 | 280 | 72 | ||
NM_001267055.1 | NP_001253984.1 | 2 - 3 | 244 | 72 | ||
XM_006238241.2 | XP_006238303.1 | 3 - 4 | 368 | 72 | ||
XM_006238242.2 | XP_006238304.1 | 2 - 3 | 256 | 72 | ||
XM_008763796.1 | XP_008762018.1 | 3 - 4 | 291 | 72 | ||
GenBank mRNA | AY156922.1 | - | 3 - 4 | 348 | 72 | |
BC086343.1 | - | 3 - 4 | 341 | 7301739 | 72 | |
DY317988.1 | - | 2 - 3 | 239 | 72 | ||
FM103539.1 | - | 2 - 3 | 180 | 72 | ||
FQ217142.1 | - | 2 - 3 | 238 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |