Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tp53inp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270947.1 | NP_001257876.1 | 3 - 4 | 662 | 75 | |
XM_003749598.3 | XP_003749646.1 | 3 - 4 | 800 | 75 | ||
XM_006235327.2 | XP_006235389.1 | 3 - 4 | 1374 | 75 | ||
XM_006235329.2 | XP_006235391.1 | 3 - 4 | 1568 | 75 | ||
GenBank mRNA | AJ297794.1 | - | 3 - 4 | 612 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |