Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd11b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017081.2 | NP_058777.1 | 3 - 4 | 793 | 89 | |
XM_006255445.2 | XP_006255507.1 | 3 - 4 | 1509 | 89 | ||
GenBank mRNA | BC087023.1 | - | 3 - 4 | 780 | 7130250 | 89 |
U22424.1 | - | 3 - 4 | 794 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |