Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053338.2 | NP_445790.2 | 4 - 5 | 822 | 90 | |
XM_006255055.2 | XP_006255117.1 | 4 - 5 | 904 | 90 | ||
GenBank mRNA | FM055705.1 | - | 1 - 2 | 163 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons |