Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133610.2 | NP_598294.1 | 6 - 7 | 1209 | 100 | |
GenBank mRNA | AF073891.1 | - | 1 - 2 | 286 | 100 | |
AF185637.1 | - | 6 - 7 | 1168 | 100 | ||
AJ250280.1 | - | 6 - 7 | 1211 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Rattus norvegicus Probe spans exons |