Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207594.2 | NP_997477.1 | 3 - 4 | 389 | 102 | |
GenBank mRNA | BC061570.1 | - | 3 - 4 | 393 | 6918143 | 102 |
FQ210595.1 | - | 3 - 4 | 367 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |