Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001180140.1 | NP_010202.1 | 1 | 144 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |