Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001180184.1 | NP_010158.1 | 1 - 2 | 95 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Saccharomyces cerevisiae Probe spans exons |