Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSP78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001180566.3 | NP_010544.3 | 1 | 2354 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |