Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZUO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001181414.1 | NP_011801.1 | 1 | 1241 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Saccharomyces cerevisiae Inventoried Both primers and probe map within a single exon |