Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184522.3 | NP_878113.4 | 1 - 2 | 32 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Saccharomyces cerevisiae Probe spans exons |