Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNISR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113439.1 | NP_001106910.1 | 9 - 10 | 1319 | 53 | |
XM_021079463.1 | XP_020935122.1 | 2071 | 53 | |||
XM_021080246.1 | XP_020935905.1 | 1250 | 53 | |||
GenBank mRNA | EU308569.1 | - | 9 - 10 | 1319 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons |