Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC42SE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190193.1 | NP_001177122.1 | 3 - 4 | 491 | 64 | |
XM_005663461.3 | XP_005663518.1 | 572 | 64 | |||
XM_013997164.2 | XP_013852618.1 | 725 | 64 | |||
XM_021088571.1 | XP_020944230.1 | 422 | 64 | |||
GenBank mRNA | AK239716.1 | - | 3 - 4 | 486 | 64 | |
BW960573.1 | - | 3 - 4 | 491 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |