Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031786.1 | NP_001026956.1 | 3 - 4 | 244 | 115 | |
XM_005657393.3 | XP_005657450.1 | 406 | 115 | |||
XM_005657394.3 | XP_005657451.1 | 434 | 115 | |||
GenBank mRNA | AK234361.1 | - | 4 - 5 | 369 | 115 | |
AK350329.1 | - | 4 - 5 | 359 | 115 | ||
AK399576.1 | - | 4 - 5 | 372 | 115 | ||
AY996812.1 | - | 3 - 4 | 244 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |