Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244132.1 | NP_001231061.1 | 3 - 4 | 915 | 63 | |
GenBank mRNA | AF178675.1 | - | 1 - 2 | 127 | 63 | |
AK347741.1 | - | 3 - 4 | 915 | 63 | ||
AK400539.1 | - | 3 - 4 | 911 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Probe spans exons |