Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130211.1 | NP_001123683.1 | 5 - 6 | 837 | 88 | |
GenBank mRNA | AF311909.1 | - | 3 - 4 | 319 | 88 | |
AK231059.1 | - | 5 - 6 | 670 | 88 | ||
AK351736.1 | - | 5 - 6 | 650 | 88 | ||
AK391541.1 | - | 5 - 6 | 652 | 88 | ||
AY923074.1 | - | 5 - 6 | 837 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |