Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005671462.3 | XP_005671519.2 | 5104 | 104 | ||
GenBank mRNA | AK348444.1 | - | 15 - 16 | 2011 | 104 | |
AY284842.1 | - | 1 - 2 | 178 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |