Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005671462.3 | XP_005671519.2 | 5463 | 60 | ||
GenBank mRNA | AK348444.1 | - | 18 - 19 | 2370 | 60 | |
AY284842.1 | - | 4 - 5 | 537 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons |