Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244168.1 | NP_001231097.1 | 1 | 765 | 56 | |
GenBank mRNA | AY691682.1 | - | 1 | 347 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |