Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS4X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204283.1 | NP_001191212.1 | 3 - 4 | 305 | 71 | |
GenBank mRNA | AK232196.1 | - | 3 - 4 | 305 | 71 | |
AK233807.1 | - | 3 - 4 | 279 | 71 | ||
AK394418.1 | - | 3 - 4 | 312 | 71 | ||
D50110.1 | - | 2 - 3 | 199 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |