Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170768.1 | NP_001164239.1 | 6 | 985 | 101 | |
GenBank mRNA | AK240474.1 | - | 6 | 974 | 101 | |
AK391478.1 | - | 6 | 101 | |||
GQ184631.1 | - | 6 | 985 | 101 | ||
GQ184632.1 | - | 6 | 985 | 101 | ||
L48205.1 | - | 3 | 450 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |