Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005660372.3 | XP_005660429.1 | 691 | 145 | ||
GenBank mRNA | AK233369.1 | - | 5 - 6 | 574 | 145 | |
AK346200.1 | - | 5 - 6 | 145 | |||
M35990.1 | - | 5 - 6 | 480 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |