Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHSG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005652369.3 | XP_005652426.1 | 856 | 85 | ||
GenBank mRNA | AK233033.1 | - | 2 - 3 | 412 | 85 | |
X56021.1 | - | 326 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |