Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNLY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278755.1 | NP_001265684.1 | 4 | 278 | 72 | |
GenBank mRNA | AK344288.1 | - | 4 | 289 | 72 | |
KC346971.1 | - | 4 | 278 | 72 | ||
KC346972.1 | - | 4 | 278 | 72 | ||
X85431.1 | - | 451 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons |