Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244131.1 | NP_001231060.1 | 2 | 410 | 72 | |
GenBank mRNA | AF288821.1 | - | 2 | 157 | 72 | |
AK233768.1 | - | 2 | 364 | 72 | ||
AK236777.1 | - | 2 | 368 | 72 | ||
AK237429.1 | - | 2 | 364 | 72 | ||
AK237544.1 | - | 2 | 364 | 72 | ||
AK238544.1 | - | 1 | 530 | 72 | ||
AK346091.1 | - | 2 | 364 | 72 | ||
AK346206.1 | - | 2 | 364 | 72 | ||
AK389759.1 | - | 2 | 315 | 72 | ||
AK392233.1 | - | 2 | 368 | 72 | ||
AK395141.1 | - | 1 | 533 | 72 | ||
AK399198.1 | - | 2 | 366 | 72 | ||
AK400010.1 | - | 2 | 366 | 72 | ||
AY610290.1 | - | 2 | 204 | 72 | ||
CX061020.1 | - | 2 | 441 | 72 | ||
FS674820.1 | - | 2 | 356 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |