Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021086531.1 | XP_020942190.1 | 2574 | 71 | ||
GenBank mRNA | AF319662.1 | - | 1253 | 71 | ||
AK391806.1 | - | 4 | 2542 | 71 | ||
AK399864.1 | - | 2 | 1412 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |