Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001861.2 | NP_001001861.1 | 4 | 949 | 133 | |
GenBank mRNA | AY577905.1 | - | 4 | 950 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Both primers and probe map within a single exon |