Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008686.1 | NP_001008686.1 | 4 | 352 | 149 | |
XM_005653705.3 | XP_005653762.1 | 448 | 149 | |||
XM_005653706.3 | XP_005653763.1 | 446 | 149 | |||
XM_005653707.3 | XP_005653764.1 | 446 | 149 | |||
GenBank mRNA | AB190229.1 | - | 4 | 352 | 149 | |
AK233822.1 | - | 4 | 365 | 149 | ||
AY823638.1 | - | 4 | 436 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons |