Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077229.1 | NP_001070697.1 | 2 | 551 | 133 | |
XM_021066262.1 | XP_020921921.1 | 816 | 133 | |||
XM_021066263.1 | XP_020921922.1 | 822 | 133 | |||
GenBank mRNA | AK235325.1 | - | 2 | 776 | 133 | |
AK390988.1 | - | 2 | 779 | 133 | ||
AK396414.1 | - | 2 | 773 | 133 | ||
AK398220.1 | - | 770 | 133 | |||
AK398822.1 | - | 2 | 775 | 133 | ||
AK398885.1 | - | 2 | 775 | 133 | ||
AK400839.1 | - | 2 | 778 | 133 | ||
DQ972966.1 | - | 2 | 551 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |