Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AWN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ238538.1 | - | 4 | 351 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Amplicon spans exons and probe does not span exons |