Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213884.1 | NP_999049.1 | 35 | 4655 | 63 | |
GenBank mRNA | AB009387.1 | - | 35 | 4655 | 63 | |
AK389812.1 | - | 16 | 2147 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |