Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHCGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214449.1 | NP_999614.1 | 11 | 2050 | 64 | |
XM_021085886.1 | XP_020941545.1 | 1965 | 64 | |||
XM_021085887.1 | XP_020941546.1 | 1334 | 64 | |||
XM_021085888.1 | XP_020941547.1 | 1107 | 64 | |||
GenBank mRNA | JN120794.1 | - | 11 | 2007 | 64 | |
JN120795.1 | - | 12 | 2177 | 64 | ||
JN120796.1 | - | 11 | 1297 | 64 | ||
JN120797.1 | - | 11 | 1066 | 64 | ||
JN120798.1 | - | 10 | 998 | 64 | ||
M29525.1 | - | 11 | 2050 | 64 | ||
M29526.1 | - | 11 | 1784 | 64 | ||
M29527.1 | - | 11 | 1340 | 64 |
Gene Symbol: LOC100623180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001315660.1 | NP_001302589.1 | 11 | 1066 | 64 | |
GenBank mRNA | M29528.1 | - | 11 | 1114 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Both primers and probe map within a single exon |