Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097470.2 | NP_001090939.1 | 2 | 153 | 65 | |
GenBank mRNA | AK347150.1 | - | 2 | 154 | 65 | |
AK391626.1 | - | 2 | 154 | 65 | ||
DQ629148.1 | - | 2 | 154 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |