Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101031.2 | NP_001094501.1 | 3 | 1349 | 68 | |
XM_005659968.2 | XP_005660025.1 | 1639 | 68 | |||
XM_005659970.3 | XP_005660027.1 | 2135 | 68 | |||
XM_021069353.1 | XP_020925012.1 | 2668 | 68 | |||
XM_021069851.1 | XP_020925510.1 | 1349 | 68 | |||
GenBank mRNA | EU026433.2 | - | 3 | 1353 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Both primers and probe map within a single exon |