Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123180.1 | NP_001116652.1 | 5 | 553 | 155 | |
XM_021068191.1 | XP_020923850.1 | 1059 | 155 | |||
XM_021068192.1 | XP_020923851.1 | 1354 | 155 | |||
GenBank mRNA | AK231680.1 | - | 6 | 985 | 155 | |
AK237022.1 | - | 6 | 970 | 154 | ||
AK237137.1 | - | 6 | 990 | 154 | ||
AK238898.1 | - | 6 | 973 | 155 | ||
AK239425.1 | - | 6 | 1003 | 155 | ||
AK350463.1 | - | 6 | 773 | 155 | ||
AK394394.1 | - | 6 | 794 | 155 | ||
AK396180.1 | - | 6 | 1280 | 155 | ||
AK399105.1 | - | 6 | 777 | 155 | ||
AK400129.1 | - | 6 | 834 | 155 | ||
AY610083.1 | - | 6 | 912 | 155 | ||
DQ917629.1 | - | 5 | 553 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons |